missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide hydrate, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11445210
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445210 25 g 25g
11455210 100 g 100g
11465210 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445210 Supplier Thermo Scientific Alfa Aesar Supplier No. 044498.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681435-25-6
Molecular Weight (g/mol) 272.22
MDL Number MFCD00150204
InChI Key KPWJBEFBFLRCLH-UHFFFAOYSA-L
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,
IUPAC Name cadmium(2+) dibromide
SMILES [Br-].[Br-].[Cd++]

Specifications

Color White to Yellow
Physical Form Crystalline
Quantity 25 g
Molecular Formula Br2Cd
UN Number UN2570
Formula Weight 272.22 (Anhydrous)
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.