missing translation for 'onlineSavingsMsg'
Learn More

Cadmium acetate dihydrate, 99.999% (metals basis)

Product Code. 11371938
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371938 100 g 100g
11351938 25 g 25g
11361938 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11371938 Supplier Thermo Scientific Alfa Aesar Supplier No. 011865.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5743-04-4 | C4H10CdO6 | 266.53 g/mol

It is used in dyestuff intermediates, ceramics and producing iridescent effects on porcelains and pottery. It acts as a reagent for determination of S, Se, and Te. It is also used in cadmium electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00012449
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in water,ethanol and methanol.
Formula Weight 266.54 (230.50 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 2.01
Chemical Name or Material Cadmium acetate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.