missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Solstice

Product Code. 15641880 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15641880 10 g Poly Bottle 10g
15631880 100 g Poly Bottle 100g
15651880 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15641880 Supplier Honeywell Research Chemicals Supplier No. 2090110G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Melting Point 256°C
Physical Form Solid
Quantity 10 g
Linear Formula Cd(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3730567
Formula Weight 266.53g/mol
Percent Purity ≥98.0% (KT)
Grade purum p.a.
Packaging Poly Bottle
Density 2.01g/cu.cm
Chemical Name or Material Cadmium Acetate Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.