missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Acetate Dihydrate, 98%, Reagent grade, Honeywell™

Product Code. p-7120270 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15632940 100 g Poly Bottle 100g
15642940 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15632940 Supplier Honeywell Research Chemicals Supplier No. 289159100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Melting Point 256°C
Physical Form Solid
Quantity 100 g
Linear Formula Cd(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3730567
Formula Weight 266.53g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Density 2.01g/cm3
Chemical Name or Material Cadmium Acetate Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.