missing translation for 'onlineSavingsMsg'
Learn More

Cadmium acetate dihydrate, 98%, pure

Product Code. 15289666 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15289666 5 kg 5kg
15249716 250 g 250g
15279666 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15289666 Supplier Thermo Scientific Acros Supplier No. 212310050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5743-04-4 | C4H6CdO4·2H2O | 266.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-04-4
Molecular Formula C4H6CdO4·2H2O
Molecular Weight (g/mol) 266.52
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L

Specifications

Quantity 5 kg
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula (CH3CO2)2Cd·2H2O
Merck Index 10,1584
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 266.52
Percent Purity 98%
Grade Pure
Loss on Drying 12.5% to 14.5%
Chemical Name or Material Cadmium acetate dihydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium acetate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.