missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CADM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549907
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total IGSF4C / CADM4 protein.
CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
Specifications
| CADM4 | |
| Polyclonal | |
| Unconjugated | |
| CADM4 | |
| Cadm4; Cell adhesion molecule 4; Igdf4c; IGSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; Necl4; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; synCAM4; TSLC1-like 2; TSLC1-like protein 2; Tsll2; Upar; urokinase plasminogen activator receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 199731, 260299, 365216 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q1WIM1, Q8NFZ8, Q8R464 | |
| CADM4 | |
| Synthesized peptide derived from internal of human CADM4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction