missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CADM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533378
This item is not returnable.
View return policy
Description
CADM4 Polyclonal Antibody for IHC (P)
CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
Specifications
| CADM4 | |
| Polyclonal | |
| Unconjugated | |
| CADM4 | |
| Cadm4; Cell adhesion molecule 4; Igdf4c; IGSF4C; Immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; Necl4; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; synCAM4; TSLC1-like 2; TSLC1-like protein 2; Tsll2; Upar; urokinase plasminogen activator receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100070257, 100351370, 100523382, 199731, 260299, 365216, 484465, 523277 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.8 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q1WIM1, Q8NFZ8, Q8R464 | |
| CADM4 | |
| Synthetic 14 amino acid peptide from internal region of human CADM4 / NECL-4. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction