missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CADM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532687
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bovine, Dog, Bat, Elephant, Panda, Horse, Rabbit, Pig, Opossum (100%).
This gene encodes a member of the synaptic cell adhesion molecule 1 (SynCAM) family which belongs to the immunoglobulin (Ig) superfamily. The encoded protein has three Ig-like domains and a cytosolic protein 4.1 binding site near the C-terminus. Proteins belonging to the protein 4.1 family crosslink spectrin and interact with other cytoskeletal proteins. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
CADM2 | |
Polyclonal | |
Unconjugated | |
CADM2 | |
2900078E11Rik; 9330131D06; A830029E02Rik; Cadm2; Cell adhesion molecule 2; Igdf4d; Igsf4d; immunoglobulin superfamily member 4D; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D; NECL3; Necl-3; nectin-like 3; nectin-like protein 3; Synaptic cell adhesion molecule 2; SynCAM 2; SynCAM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100069765, 100340168, 100626294, 253559, 487680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8N3J6 | |
CADM2 | |
Synthetic 16 amino acid peptide from extracellular domain of human CADM2. | |
50 μg | |
Primary | |
Canine, Equine, Human, Non-human Primate, Porcine, Rabbit | |
Antibody | |
IgG |