Learn More
Invitrogen™ CADM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533377
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Galago, Marmoset, Mouse, Rat, Hamster, Elephant, Panda, Bovine, Dog, Bat, Horse, Rabbit, Pig, Opossum, Guinea pig, Zebra finch, Platypus, Xenopus (100%); Turkey, Chicken (94%); Zebrafish (82%). Antigen retrieval with HIER and sodium citrate at pH 6.0 is required for Immunohistochemistry using formalin-fixed paraffin-embedded human tissues.
This gene encodes a member of the synaptic cell adhesion molecule 1 (SynCAM) family which belongs to the immunoglobulin (Ig) superfamily. The encoded protein has three Ig-like domains and a cytosolic protein 4.1 binding site near the C-terminus. Proteins belonging to the protein 4.1 family crosslink spectrin and interact with other cytoskeletal proteins. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CADM2 | |
| Polyclonal | |
| Unconjugated | |
| CADM2 | |
| 2900078E11Rik; 9330131D06; A830029E02Rik; Cadm2; Cell adhesion molecule 2; Igdf4d; Igsf4d; immunoglobulin superfamily member 4D; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D; NECL3; Necl-3; nectin-like 3; nectin-like protein 3; Synaptic cell adhesion molecule 2; SynCAM 2; SynCAM2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100069765, 100340168, 100626294, 100734249, 100773220, 239857, 253559, 360687, 487680, 517759 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q1WIM2, Q8BLQ9, Q8N3J6 | |
| CADM2 | |
| Synthetic 17 amino acid peptide from internal region of human CADM2. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Guinea Pig, Frog, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.