missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CADM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106550
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CADM2.
This gene encodes a member of the synaptic cell adhesion molecule 1 (SynCAM) family which belongs to the immunoglobulin (Ig) superfamily. The encoded protein has three Ig-like domains and a cytosolic protein 4.1 binding site near the C-terminus. Proteins belonging to the protein 4.1 family crosslink spectrin and interact with other cytoskeletal proteins. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CADM2 | |
| Polyclonal | |
| Unconjugated | |
| CADM2 | |
| 2900078E11Rik; 9330131D06; A830029E02Rik; Cadm2; Cell adhesion molecule 2; Igdf4d; Igsf4d; immunoglobulin superfamily member 4D; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D; NECL3; Necl-3; nectin-like 3; nectin-like protein 3; Synaptic cell adhesion molecule 2; SynCAM 2; SynCAM2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 239857, 253559, 360687 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q1WIM2, Q8BLQ9, Q8N3J6 | |
| CADM2 | |
| A synthesized peptide derived from human CADM2(Accession Q8N3J6), corresponding to amino acid residues E138-L188. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction