missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cadherin E Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cadherin E Ab-4, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1862-S1

 View more versions of this product

Product Code. 12640347

  • ONLINE EXCLUSIVE PRICE
    £424.00
    £397.77 / 500µL
    Save £26.23
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.

Host Species: Mouse

Clone: NCH-38

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: E-cadherin and GST recombinant protein

Molecular Weight: 120 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: IVD; Int'l: IVD

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cadherin E/E-Cadherin Ab-4
Monoclonal
Unconjugated
E-cadherin and GST recombinant protein
500 μL
Junction Protein
Human
IgG2a κ
Immunohistochemistry (Paraffin)
NCH-38
Mouse
Breast CA
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.