missing translation for 'onlineSavingsMsg'
Learn More

Cadherin-7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16821894 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16821894 20 μL 20µL
16811894 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16821894 Supplier Proteintech Supplier No. 135981AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene is a type II classical cadherin from the cadherin superfamily. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Cadherins mediate cell-cell binding in a homophilic manner, contributing to the sorting of heterogeneous cell types and the maintece of orderly structures. Alternative splicing in the 5′ UTR of this gene yields variant transcripts encoding the same protein.

Specifications

Antigen Cadherin-7
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDH7
Gene Accession No. Q5DWV2, Q8BM92, Q9ULB5
Gene Alias cadherin 7, type 2, Cadherin-7, CDH7, CDH7L1
Gene Symbols Cdh7
Host Species Rabbit
Immunogen Cadherin-7 Fusion Protein Ag4518
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1005, 241201, 29162
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.