missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13598-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a type II classical cadherin from the cadherin superfamily. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Cadherins mediate cell-cell binding in a homophilic manner, contributing to the sorting of heterogeneous cell types and the maintece of orderly structures. Alternative splicing in the 5′ UTR of this gene yields variant transcripts encoding the same protein.Specifications
| Cadherin-7 | |
| Polyclonal | |
| Unconjugated | |
| CDH7 | |
| cadherin 7, type 2, Cadherin-7, CDH7, CDH7L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1005, 241201, 29162 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5DWV2, Q8BM92, Q9ULB5 | |
| Cdh7 | |
| Cadherin-7 Fusion Protein Ag4518 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction