missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cadherin-6/KCAD 293T Cell Transient Overexpression Lysate

Product Code. 18367777 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367777 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367777 Supplier Novus Biologicals™ Supplier No. H00001004T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Se returpolicy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifikationer

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Research Category Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
Target Cadherin-6/KCAD
Synonym cadherin 6, type 2, K-cadherin (fetal kidney), cadherin, fetal kidney, cadherin-6, KCAD, K-cadherin, K-cadherin (fetal kidney)
Gene Symbol CDH6
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.