missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-4/R-Cadherin Antibody (OTI4F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46208
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Cadherin-4/R-Cadherin Monoclonal specifically detects Cadherin-4/R-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cadherin-4/R-Cadherin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
| Mouse | |
| 98.3 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI4F5 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| CDH4 | |
| Human recombinant protein fragment corresponding to amino acids 170-428 of human CDH4 (NP_001785) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 1002 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction