missing translation for 'onlineSavingsMsg'
Learn More

Cadherin-26 Antibody, Novus Biologicals™

Codice prodotto. 18419290 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantity:
0.1 mL
25ul
Dimensione della confezione:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18419290 25ul 25µL
18270806 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18419290 Fornitore Novus Biologicals N. del fornitore NBP18062325ul

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

Cadherin-26 Polyclonal specifically detects Cadherin-26 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifica

Antigen Cadherin-26
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias cadherin 26, cadherin-like 26, cadherin-like protein 26, Cadherin-like protein VR20, VR20
Gene Symbols CDH26
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:VIIIHAVDDGFPPQTATGTLMLFLSDINDNVPTLRPRSRYMEVCESAVHEPLHIEAEDPDLEPFSDPFTFELDNTWGNAEDTWKLGRNWGQSVELLTLRSLPRGNYLVPLFIGDKQGLSQKQTVHVRICPCASGLTCVELADAEVGL
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60437
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Vedi altri risultati Mostra meno risultati

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.