missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-23 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92450-0.02ml
This item is not returnable.
View return policy
Description
Cadherin-23 Polyclonal antibody specifically detects Cadherin-23 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Cadherin-23 | |
| Polyclonal | |
| Western Blot 1:200-1:1000 | |
| cadherin related 23, cadherin-23, cadherin-like 23, cadherin-related 23, cadherin-related family member 23, CDHR23, DKFZp434P2350, FLJ00233, FLJ36499, KIAA1774, KIAA1812, MGC102761, Otocadherin, USH1D | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-270 of human CDH23 (NP_001165402.1). LLISEDTPVGSSVTQLLAQDMDNDPLVFGVSGEEASRFFAVEPDTGVVWLRQPLDRETKSEFTVEFSVSDHQGVITRKVNIQVGDVNDNAPTFHNQPYSVRIPENTPVGTPIFIVNATDPDLGAGGSVLYSFQPPSQFFAIDSARGIVTVIRELDYETTQAYQLTVNATDQDKTRPLSTLANLAIIITDVQDMDPIFINLPYSTNIYEHSPPGTTVRIITAIDQDKGRPRG | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 64072 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction