missing translation for 'onlineSavingsMsg'
Learn More

Cadherin-17 Antibody (029), HRP, Novus Biologicals™

Product Code. 18320770 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320770 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320770 Supplier Novus Biologicals Supplier No. NBP289891H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cadherin-17 Monoclonal specifically detects Cadherin-17 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cadherin-17
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 29
Conjugate HRP
Formulation PBS
Gene Alias cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Cadherin-17 (Q12864-1; Met1-Met787).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1015
Test Specificity No cross-reactivity in ELISA with: Human Cad8 Human E-Cad Human Cad5
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.