missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-16 Mouse anti-Human, Mouse, Rat, Canine, Rabbit, Clone: rCDH16/1071, Novus Biologicals™
Mouse Monoclonal Antibody
£406.00
Specifications
| Antigen | Cadherin-16 |
|---|---|
| Clone | rCDH16/1071 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
Cadherin-16 Monoclonal specifically detects Cadherin-16 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
| Cadherin-16 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Canine, Rabbit | |
| cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
| Recombinant full-length human Cadherin-16 protein | |
| Primary | |
| Store at 4°C. |
| rCDH16/1071 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1014 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title