missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-16 Mouse anti-Human, Clone: CDH16/2448, Novus Biologicals™
Mouse Monoclonal Antibody
£205.00
Specifications
| Antigen | Cadherin-16 |
|---|---|
| Clone | CDH16/2448 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Description
Cadherin-16 Monoclonal specifically detects Cadherin-16 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cadherin-16 | |
| 0.2 mg/ml | |
| ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
| Recombinant fragment (around aa 371-507) of human Cadherin-16 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| CDH16/2448 | |
| ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1014 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title