missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-16, Mouse anti-Human, Clone: CDH16/2125, Novus Biologicals™
Mouse Monoclonal Antibody
£203.00 - £377.00
Specifications
| Antigen | Cadherin-16 |
|---|---|
| Clone | CDH16/2125 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18275988
|
Novus Biologicals
NBP2-79722-20UG |
20 μg |
£203.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18255998
|
Novus Biologicals
NBP2-79722-100UG |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadherin-16 Monoclonal specifically detects Cadherin-16 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cadherin-16 | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75309 | |
| 1014 | |
| Recombinant fragment (around aa 371-507) of human Cadherin-16 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| CDH16/2125 | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin | |
| CDH16 | |
| IgG1 κ | |
| Protein A or G purified | |
| This monoclonal antibody recognizes a protein of 130kDa, identified as Cadherin-16; Cadherin-16 has been suggested to distinguish Chromophobe Renal-Cell Carcinoma from Oncocytoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title