missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12618-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3′ trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| Cadherin-13 | |
| Polyclonal | |
| Unconjugated | |
| CDH13 | |
| Cadherin 13, Cadherin-13, CDH13, CDHH, H cadherin, H-cadherin, Heart cadherin, P105, T cad, T cadherin, T-cadherin, Truncated cadherin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1012, 12554 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55290, Q9WTR5 | |
| CDH13 | |
| Cadherin-13 Fusion Protein Ag3295 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction