missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-13 Antibody (CL15368) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
£285.00 - £426.00
Specifications
| Antigen | Cadherin-13 |
|---|---|
| Clone | CL15368 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:1000-1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279743
|
Novus Biologicals
NBP3-43666-25ul |
25 μL |
£285.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279475
|
Novus Biologicals
NBP3-43666-100ul |
100 μL |
£426.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadherin-13 Monoclonal antibody specifically detects Cadherin-13 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| Cadherin-13 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS, pH 7.2, 40% glycerol | |
| 1012 | |
| IgG1 | |
| Protein A purified |
| CL15368 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000-1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cadherin 13, H-cadherin (heart), cadherin-13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, Truncated cadherin, truncated-cadherin | |
| This antibody was generated using a recombinant protein sequence of P55290, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title