missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-12 Antibody (343621), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2240W
This item is not returnable.
View return policy
Description
Cadherin-12 Monoclonal antibody specifically detects Cadherin-12 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Cadherin-12 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1010 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 343621 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| BR-cadherin, cadherin 12, type 2 (N-cadherin 2), cadherin-12, FLJ34857, N-cadherin 2, Neural type cadherin 2, neural type, 2, neuronal cadherin 2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-12, Met1-Pro609, Accession # P55289 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction