missing translation for 'onlineSavingsMsg'
Learn More

Cadherin-11 Antibody (283416) [DyLight 550], Novus Biologicals™

Product Code. 30736696 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736696 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736696 Supplier Novus Biologicals Supplier No. FAB1790L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cadherin-11 Monoclonal antibody specifically detects Cadherin-11 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cadherin-11
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 283416
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, CDHOB, OB, OB-cadherin, Osteoblast cadherin
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cadherin-11, Phe23-Thr617, Accession # AAA35622
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1009
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.