missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAD Recombinant Rabbit Monoclonal Antibody (23GB3075), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen CAD
Clone 23GB3075
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089684
GREENER_CHOICE
View Documents
Invitrogen™
MA552604
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089014
View Documents
Invitrogen™
MA552605
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CAD Recombinant Monoclonal antibody specifically detects CAD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

CAD
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
P27708
790
IgG
Affinity chromatography
Antibody
23GB3075
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
2410008J01Rik; 38E.19; anon-WO2004063362.83; Aspartate carbamoyltransferase; AU018859; cad; CAD protein; CAD trifunctional protein; cad-PA; cad-PB; cad-PC; carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase; carbamoylphosphate synthetase 2/aspartate transcarbamylase/dihydroorotase; carbamyl phosphatate synthetase 2; caudal; cb456; cd; CDG1Z; CG1759; CG1759-PA; CG1759-PB; CG1759-PC; Cpad; Dihydroorotase; Dmel\CG1759; Dmel_CG1759; Glutamine-dependent carbamoyl-phosphate synthase; Homeotic protein caudal; LD29596p; multifunctional protein CAD; S67; si:dkey-221h15.3; wu:fc30c12; wu:fc33d01; wu:fc67g02
A synthetic peptide of human CAD (1-34AA).
Primary
-20°C
CAD
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.