missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACYBP Antibody (7D11A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27912
This item is not returnable.
View return policy
Description
CACYBP Monoclonal antibody specifically detects CACYBP in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CACYBP | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| CacyBP, calcyclin binding protein, GIG5, hCacyBP, MGC87971, RP1-102G20.6, S100A6-binding protein, S100A6BPgrowth-inhibiting gene 5 protein, Siah-interacting protein, Siah-interacting protein (SIP), SIPcalcyclin-binding protein | |
| Truncated recombinant CACYBP expressed in E. Coli strain BL21 (DE3). | |
| 0.1 mg | |
| Wnt Signaling Pathway | |
| 27101 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| 7D11A8 | |
| Western Blot 1:200 - 1:1000, ELISA 1:10, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| Q9HB71 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction