missing translation for 'onlineSavingsMsg'
Learn More

Cacodylic Acid, TRC

Product Code. 30338084
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30338084 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30338084

Brand: Toronto Research Chemicals TRCC05250010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cacodylic acid
CAS 75-60-5
Molecular Formula C2 H7 As O2
Quantity 10 g
Synonym Arsine oxide, hydroxydimethyl- (7CI,8CI), Arsinic acid, dimethyl- (9CI), Cacodylic acid (6CI), As,As-Dimethylarsinic acid, Ansar 138, Arsan, Dimethylarsenic acid, Dimethylarsinic acid, Hydroxydimethylarsine oxide, NSC 103115, NSC 71157, NSC 71158, Phytar, Silvisar 510, Sylvicor
InChI Formula InChI=1S/C2H7AsO2/c1-3(2,4)5/h1-2H3,(H,4,5)
SMILES C[As](=O)(C)O
IUPAC Name dimethylarsinic acid
Molecular Weight (g/mol) 138
Formula Weight 137.9662
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.