Learn More
Invitrogen™ CACNG8 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5114707
Product Code. 17227473
  • £455.00 / 100µL
Description
Antibody detects endogenous levels of total CACNG8.
Regulates the trafficking and gating properties of AMPA- selective glutamate receptors (AMPARs). Promotes their targeting to the cell membrane and synapses and modulates their gating properties by slowing their rates of activation, deactivation and desensitization and by mediating their resensitization. Does not show subunit-specific AMPA receptor regulation and regulates all AMPAR subunits. Thought to stabilize the calcium channel in an inactivated (closed) state.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CACNG8
Polyclonal
Unconjugated
CACNG8
CACNG6; Cacng8; calcium channel, voltage-dependent, gamma subunit 8; calcium voltage-gated channel auxiliary subunit gamma 8; CaVGamma8; mir-935; Neuronal voltage-gated calcium channel gamma-8 subunit; TARP gamma 8; TARP gamma-8; Transmembrane AMPAR regulatory protein gamma-8; voltage-dependent calcium channel gamma-8 subunit
Rabbit
Affinity chromatography
RUO
140729, 59283, 81905
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
Q8VHW2, Q8VHW5, Q8WXS5
CACNG8
A synthesized peptide derived from human CACNG8(Accession Q8WXS5), corresponding to amino acid residues V167-E217.
100 μL
Primary
Human, Mouse, Rat