missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CACNG7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106769
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CACNG7.
The protein encoded by this gene is a type II transmembrane AMPA receptor regulatory protein (TARP). TARPs regulate both trafficking and channel gating of the AMPA receptors. This gene is part of a functionally diverse eight-member protein subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two family members, a type I TARP and a calcium channel gamma subunit.
Specifications
| CACNG7 | |
| Polyclonal | |
| Unconjugated | |
| Cacng7 | |
| Cacng7; calcium channel, voltage-dependent, gamma subunit 7; calcium voltage-gated channel auxiliary subunit gamma 7; neuronal voltage-gated calcium channel gamma-7 subunit; TARP gamma-7; transmembrane AMPAR regulatory protein gamma-7; voltage dependent calcium channel gamma 7 subunit; voltage-dependent calcium channel gamma-7 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140728, 59284, 81904 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62955, P62956, P62957 | |
| Cacng7 | |
| A synthesized peptide derived from human CACNG7(Accession P62955), corresponding to amino acid residues Y198-S248. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction