missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG6 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577348
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Voltage-dependent calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of two known gamma subunit proteins. This particular gamma subunit is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is part of a functionally diverse eight-member protein subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two family members that function as transmembrane AMPA receptor regulatory proteins (TARPs). Alternative splicing results in multiple transcript variants. Variants in this gene have been associated with aspirin-intolerant asthma.
Specifications
| CACNG6 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| Cacng6 | |
| CaVgamma6 (extracellular) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140727, 54378 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.85 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q8VHW3, Q8VHW7 | |
| Cacng6 | |
| Peptide CIKRLWQADVPAGR, corresponding to amino acid residues 87-100 of rat Cavgamma6 | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction