missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CACNG5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568377
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
L-type calcium channels are composed of five subunits. The protein Calcium Channel 5 represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state.
Specifications
| CACNG5 | |
| Polyclonal | |
| Unconjugated | |
| Cacng5 | |
| Cacng5; calcium channel, voltage-dependent, gamma subunit 5; calcium voltage-gated channel auxiliary subunit gamma 5; neuronal voltage-gated calcium channel gamma-5 subunit; TARP gamma-5; transmembrane AMPAR regulatory protein gamma-5; voltage dependent calcium channel gamma 5 subunit; voltage-dependent calcium channel gamma-5 subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140723, 140726, 27091 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8VHW4, Q8VHW8, Q9UF02 | |
| Cacng5 | |
| Synthesized peptide derived from human Calcium Channel 5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction