missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CACNG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596728
This item is not returnable.
View return policy
Description
Immunogen sequence: EKNKELRFKT KREFLKASSS SPYARMPSYR YRRRRSRSSS RSTEASPSRD VSPMGLKITG AIPMGELSMY TLSREPLKVT TAASYSPDQE ASFLQVHDFF QQDLKEGFHV SMLNRRTTPV; Positive Samples: Mouse brain; Cellular Location: Cell membrane, Multi-pass membrane protein.
The expression of multiple classes of voltage dependent calcium channels (VDCC's) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC's exist before birth which is consistent with a role in migration, most N-Type VDCC's subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Specifications
| CACNG4 | |
| Polyclonal | |
| Unconjugated | |
| Cacng4 | |
| AI413107; AW491861; CACNG4; calcium channel, voltage-dependent, gamma subunit 4; calcium voltage-gated channel auxiliary subunit gamma 4; CaV gamma4; MGC11138; MGC24983; neuronal voltage-gated calcium channel gamma-4 subunit; TARP gamma-4; transmembrane AMPAR regulatory protein gamma-4; Voltage-dependent calcium channel gamma-4 subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54377 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JJV4 | |
| Cacng4 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction