Learn More
Invitrogen™ CACNG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587564
Description
Immunogen sequence: DHWAVLSPHM EHHNTTCEAA HFGLWRICTK RIPMDDSKTC GPITLPGEKN CSYFRHFNPG ESSEIFEFTT QKEYSISAAA I; Positive Samples: Mouse skeletal muscle, Mouse liver, Mouse kidney, Rat skeletal muscle, Rat liver, Rat kidney; Cellular Location: Membrane, Multi-pass membrane protein.
Voltage-dependent calcium channels are essential for the release of neurotransmitters. L-type (long lasting current) voltage-dependent calcium channels are composed of four subunits: an alpha1 subunit, a beta subunit, a gamma subunit and an alpha2 delta subunit. The beta subunit is encoded by four genes, designated beta1-beta4, all of which contribute to the diversity of calcium currents and are involved in membrane trafficking of the alpha1 subunit. L-type Ca ++ CP gamma1, also known as CACNLG or CACNG1, is a 222 amino acid multi-pass membrane protein belonging to the PMP-22/EMP/MP20 family. Expressed in skeletal muscle, L-type Ca ++ CP gamma1 is a subunit of the dihydropyridine (DHP) sensitive calcium channel and may play a role in excitation-contraction coupling. L-type Ca++ CP gamma1 is considered a novel marker for malignant hyperthermia susceptibility (MHS), an autosomal dominant disorder of skeletal muscle which manifests as a life-threatening hypermetabolic crisis triggered by commonly used inhalation anesthetics and depolarizing muscle relaxants.
Specifications
| CACNG1 | |
| Polyclonal | |
| Unconjugated | |
| Cacng1 | |
| Cacng; Cacng1; CACNLG; calcium channel, voltage-dependent, gamma subunit 1; calcium voltage-gated channel auxiliary subunit gamma 1; dihydropyridine receptor gamma 1; dihydropyridine-sensitive L-type, skeletal muscle calcium channel subunit gamma; L-type calcium channel gamma polypeptide; neuronal dihydropyridine-sensitive calcium channel gamma subunit; voltage-dependent calcium channel gamma-1 subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12299, 29658 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70578, P97707 | |
| Cacng1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.