missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35094-20ul
This item is not returnable.
View return policy
Description
CACNG1 Polyclonal antibody specifically detects CACNG1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CACNG1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:4000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 - 1:500 | |
| CACNLGDihydropyridine-sensitive L-type, skeletal muscle calcium channel subunit gamma, calcium channel, voltage-dependent, gamma subunit 1, L-type calcium channel gamma polypeptide, neuronal dihydropyridine-sensitive calcium channel gamma subunit, voltage-dependent calcium channel gamma-1 subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-110 of human CACNG1 (NP_000718.1).,, Sequence:, DHWAVLSPHMEHHNTTCEAAHFGLWRICTKRIPMDDSKTCGPITLPGEKNCSYFRHFNPGESSEIFEFTTQKEYSISAAAI | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 786 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction