missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CACNB3 Polyclonal Antibody

Description
Immunogen sequence: VYWRATHHPA PGPGLLGPPS AIPGLQNQQL LGERGEEHSP LERDSLMPSD EASESSRQAW TGSSQRSSRH LEEDYADAYQ DLYQPHRQHT SGLPSANGHD PQDRLLAQDS EHNHSDRNWQ RNRPWPKDSY; Positive Samples: Mouse brain, Rat brain.
CACNB3 encodes a regulatory beta subunit of the voltage-dependent calcium channel. Beta subunits are composed of five domains, which contribute to the regulation of surface expression and gating of calcium channels and may also play a role in the regulation of transcription factors and calcium transport.
Specifications
Specifications
| Antigen | CACNB3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.03 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Cacnb3 |
| Gene Accession No. | P54285, P54287 |
| Gene Alias | Beta3; Ca(v)beta3; CAB3; CACH3B; Cacnb3; Cacnlb3; Calcium channel subunit beta 3; calcium channel voltage-dependent subunit beta 3; calcium channel, voltage-dependent, beta 3 subunit; calcium voltage-gated channel auxiliary subunit beta 3; cardiac calcium channel beta-subunit CaB3; Cav; CCHB3; voltage-dependent calcium channel beta3A subunit; voltage-dependent L-type calcium channel subunit beta-3 |
| Gene Symbols | Cacnb3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?