missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35459-20ul
This item is not returnable.
View return policy
Description
CACNB3 Polyclonal antibody specifically detects CACNB3 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CACNB3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CAB3, CACNLB3, Calcium channel voltage-dependent subunit beta 3, calcium channel, voltage-dependent, beta 3 subunit, voltage-dependent L-type calcium channel subunit beta-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 355-484 of human CACNB3 (NP_000716.2).,, Sequence:, VYWRATHHPAPGPGLLGPPSAIPGLQNQQLLGERGEEHSPLERDSLMPSDEASESSRQAWTGSSQRSSRHLEEDYADAYQDLYQPHRQHTSGLPSANGHDPQDRLLAQDSEHNHSDRNWQRNRPWPKDSY | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 784 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction