missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNB1 Antibody (OTI3H6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | CACNB1 |
|---|---|
| Clone | OTI3H6 |
| Dilution | Western Blot 1:500-2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306686
|
Novus Biologicals
NBP3-44945-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306180
|
Novus Biologicals
NBP3-44945-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNB1 Monoclonal antibody specifically detects CACNB1 in Human,Mouse samples. It is validated for Western BlotSpecifications
| CACNB1 | |
| Western Blot 1:500-2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 782 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI3H6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| CAB1, CACNLB1calcium channel, L type, beta 1 polypeptide, Calcium channel voltage-dependent subunit beta 1, calcium channel, voltage-dependent, beta 1 subunit, CCHLB1, dihydropyridine-sensitive L-type, calcium channel beta-1 subunit, MGC41896, voltage-dependent L-type calcium channel subunit beta-1 | |
| Human recombinant protein fragment corresponding to amino acids 1-269 of human CACNB1(NP_954855) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title