Learn More
Novus Biologicals™ CACNB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30830-100ug
 View more versions of this product
Product Code. 30067150
  • £386.00 / 100µg
Description
CACNB1 Polyclonal antibody specifically detects CACNB1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CACNB1
Polyclonal
Western Blot : 5-10 μg/mL
CAB1, CACNLB1calcium channel, L type, beta 1 polypeptide, Calcium channel voltage-dependent subunit beta 1, calcium channel, voltage-dependent, beta 1 subunit, CCHLB1, dihydropyridine-sensitive L-type, calcium channel beta-1 subunit, MGC41896, voltage-dependent L-type calcium channel subunit beta-1
Synthetic peptide derived from the rat CACNB1 conjugated to KLH
100 μg
Primary
Human
Purified
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
782
Store at -20°C. Avoid freeze/thaw cycles.