missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D4 Polyclonal Antibody

CACNA2D4 Polyclonal Antibody
Brand: Invitrogen PA570453
This item is not returnable.
View return policy
Description
CACNA2D4 Polyclonal Antibody for Western Blot
Specifications
| CACNA2D4 | |
| Polyclonal | |
| Unconjugated | |
| Cacna2d4 | |
| 5730412N02Rik; BE686333; Cacna2d4; calcium channel, voltage-dependent, alpha 2/delta subunit 4; calcium voltage-gated channel auxiliary subunit alpha2delta 4; LOW QUALITY PROTEIN: voltage-dependent calcium channel subunit alpha-2/delta-4; RCD4; RGD1562038; Voltage-dependent calcium channel subunit alpha-2/delta-4; Voltage-dependent calcium channel subunit alpha-2-4; Voltage-dependent calcium channel subunit delta-4; voltage-gated calcium channel alpha(2)delta-4 subunit; voltage-gated calcium channel subunit alpha-2/delta-4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 319734 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5RJF7 | |
| Cacna2d4 | |
| synthetic peptide directed towards the middle region of human CACNA2D4 (aa 313-362). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction