missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92074-0.1ml
This item is not returnable.
View return policy
Description
CACNA2D4 Polyclonal antibody specifically detects CACNA2D4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CACNA2D4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| calcium channel, voltage-dependent, alpha 2/delta subunit 4, RCD4, voltage-dependent calcium channel subunit alpha-2/delta-4, voltage-gated calcium channel alpha(2)delta-4 subunit, Voltage-gated calcium channel subunit alpha-2/delta-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 992-1115 of human CACNA2D4 (NP_758952.4). AKSVFHHSHKHKKQDPLQPCDTEYPVFVYQPAIREANGIVECGPCQKVFVVQQIPNSNLLLLVTDPTCDCSIFPPVLQEATEVKYNASVKCDRMRSQKLRRRPDSCHAFHPEENAQDCGGASDT | |
| 0.1 mL | |
| Vision | |
| 93589 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction