missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CACNA2D4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615500
|
Novus Biologicals
NBP2-92067-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683111
|
Novus Biologicals
NBP2-92067-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNA2D4 Polyclonal antibody specifically detects CACNA2D4 in Human samples. It is validated for Western BlotSpecifications
| CACNA2D4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS with 50% glycerol, pH7.3. | |
| 93589 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| calcium channel, voltage-dependent, alpha 2/delta subunit 4, RCD4, voltage-dependent calcium channel subunit alpha-2/delta-4, voltage-gated calcium channel alpha(2)delta-4 subunit, Voltage-gated calcium channel subunit alpha-2/delta-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 992-1115 of human CACNA2D4 (NP_758952.4). AKSVFHHSHKHKKQDPLQPCDTEYPVFVYQPAIREANGIVECGPCQKVFVVQQIPNSNLLLLVTDPTCDCSIFPPVLQEATEVKYNASVKCDRMRSQKLRRRPDSCHAFHPEENAQDCGGASDT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title