missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21711-1-AP-20UL
This item is not returnable.
View return policy
Description
CACNA2D3 is a member of the alpha-2/delta subunit family, a protein in the voltage-dependent calcium channel complex. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. Research on a highly similar protein in rabbit suggests the protein described in this record is cleaved into alpha-2 and delta subunits.Specifications
| CACNA2D3 | |
| Polyclonal | |
| Unconjugated | |
| CACNA2D3 | |
| CACNA2D3, HSA272268 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12294, 55799 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IZS8, Q9Z1L5 | |
| CACNA2D3 | |
| CACNA2D3 Fusion Protein Ag16337 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction