Learn More
Invitrogen™ CACNA2D1 Monoclonal Antibody (20A)

Description
MA3-921 detects 1,4-dihydropyridine (DHP) receptor alpha-2 subunit from human, rat, mouse, guinea pig and rabbit skeletal and cardiac muscle. MA3-921 has been successfully used in Western blot, immunocytochemistry, immunofluorescence, and immunohistochemistry procedures. By Western blot, this antibody detects a 220 kDa protein under non-reducing and a 143 kDa protein under reducing conditions representing DHP from rabbit skeletal muscle membrane preparations. Immunohistochemical staining of DHP in rabbit skeletal muscle with MA3-921 results in double rows of discrete punctate staining representing pairs of triads on the opposing sides of the Z-lines. The MA3-921 antigen is purified rabbit DHP receptor.
Specifications
Specifications
| Antigen | CACNA2D1 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 20A |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.05% sodium azide |
| Gene | Cacna2d1 |
| Gene Accession No. | O08532, P13806, P54289, P54290 |
| Gene Alias | Ca(v)alpha2delta1; Cacna2; Cacna2d1; CACNL2A; Calcium channel subunit alpha 2 delta (dihydropyridine - sensitive L-type); calcium channel, L type, alpha 2 polypeptide; calcium channel, voltage-dependent, alpha 2/delta subunit 1; calcium channel, voltage-dependent, alpha2/delta subunit 1; calcium voltage-gated channel auxiliary subunit alpha2delta 1; CCHL2A; CCHLA2; DHP; DHSCCA; dihydropryridine-sensitive calcium channel alpha-2 subunit; dihydropyridine-sensitive calcium channel alpha 2; dihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit; H_DJ0560O14.1; LINC01112; lncRNA-N3; MHS3; unknown protein; voltage-dependent calcium channel alpha-2 delta subunit precursor; Voltage-dependent calcium channel subunit alpha-2/delta-1; Voltage-dependent calcium channel subunit alpha-2-1; Voltage-dependent calcium channel subunit delta-1; Voltage-gated calcium channel subunit alpha-2/delta-1; WUGSC:H_DJ0560O14.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.