missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CACNA2D1 Polyclonal antibody specifically detects CACNA2D1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | CACNA2D1 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | CACNA2, CACNL2Adihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit, calcium channel, L type, alpha 2 polypeptide, calcium channel, voltage-dependent, alpha 2/delta subunit 1, CCHL2A, MHS3, voltage-dependent calcium channel subunit alpha-2/delta-1, Voltage-gated calcium channel subunit alpha-2/delta-1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: QPKNPKSQEPVTLDFLDAELENDIKVEIRNKMIDGESGEKTFRTLVKSQDERYIDKGNRTYTWTPVNGTDYSLALVLPTYSFYYIKAKLEETITQARSKKGKMKDSETLKPDNFEESGYTFIAPRDYCNDLKISDNNTEFL |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?