missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1S Antibody (1A), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-542AF750
This item is not returnable.
View return policy
Description
CACNA1S Monoclonal antibody specifically detects CACNA1S in Human, Mouse, Rat, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| CACNA1S | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Guinea Pig, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1A | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CACH1, CACN1, CACNL1A3MHS5, calcium channel, L type, alpha 1 polypeptide, isoform 3 (skeletal muscle, Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle, calcium channel, voltage-dependent, L type, alpha 1S subunit, Cav1.1, CCHL1A3, dihydropyridine receptor, dihydropyridine-sensitive L-type calcium channel alpha-1 subunit, HOKPP, HOKPP1, hypokalemic periodic paralysis), hypoPP, TTPP1, voltage-dependent L-type calcium channel subunit alpha-1S, Voltage-gated calcium channel subunit alpha Cav1.1 | |
| Purified rabbit muscle T-tubule dihydropyridine receptor. | |
| 0.2 mL | |
| Neuroscience, Signal Transduction | |
| 779 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction