missing translation for 'onlineSavingsMsg'
Learn More

CACNA1G Antibody (S178A-9), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18410303 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410303 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410303 Supplier Novus Biologicals Supplier No. NBP259322JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CACNA1G Monoclonal antibody specifically detects CACNA1G in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA1G
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone S178A-9
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit
Host Species Mouse
Immunogen Fusion protein amino acids 2052-2172 (cytoplasmic C-terminus) of mouse Cav3.1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8913
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.