missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1G Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-45239
This item is not returnable.
View return policy
Description
CACNA1G Polyclonal specifically detects CACNA1G in Human, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.
Specifications
| CACNA1G | |
| Polyclonal | |
| Peptide ELISA Detection limit 1:32000 | |
| NP_061496.2 | |
| CACNA1G | |
| Peptide with sequence C-AEEISKREDASGQLS corresponding to internal region according to NP_061496.2, NP_938192.1, NP_938194.1, NP_938198.1, NP_938199.1, NP_938200.1, NP_938407.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_061496.2), 4 (NP_938199.1), 5 (NP_938194.1), 8 (NP_938198.1), 9 (NP_938192.1), 10 (NP_938200.1) and isoform 15 (NP_938407.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| calcium channel, voltage-dependent, T type, alpha 1G subunit, cav3.1c, KIAA1123, voltage-dependent calcium channel alpha 1G subunit, voltage-dependent T-type calcium channel subunit alpha-1G, voltage-dependent, alpha 1G subunit, voltage-dependent, T type, alpha-1G subunit | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8913 | |
| Human, Rat, Bovine, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction