missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1E Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27225-1-AP-150UL
This item is not returnable.
View return policy
Description
The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.Specifications
| CACNA1E | |
| Polyclonal | |
| Unconjugated | |
| CACNA1E | |
| BII, Brain calcium channel II, CACH6, CACNA1E, CACNL1A6, Cav2.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12290, 777 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15878, Q61290 | |
| CACNA1E | |
| CACNA1E Fusion Protein Ag25944 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction