missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1E Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | CACNA1E |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617511
|
Novus Biologicals
NBP2-92004-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605200
|
Novus Biologicals
NBP2-92004-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNA1E Polyclonal antibody specifically detects CACNA1E in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CACNA1E | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 777 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Brain calcium channel II, CACH6calcium channel, R type, alpha-1 polypeptide, CACNL1A6calcium channel, voltage-dependent, alpha 1E subunit, calcium channel, voltage-dependent, R type, alpha 1E subunit, Cav2.3, L type, alpha-1 polypeptide, L type, alpha-1 polypeptide, isoform 6, voltage-dependent calcium channel alpha 1E subunit, voltage-dependent R-type calcium channel subunit alpha-1E, voltage-gated calcium channel alpha 1E subunit, Voltage-gated calcium channel subunit alpha Cav2.3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2014-2313 of human CACNA1E (NP_001192222.1). PSSMRRSFSTIRDKRSNSSWLEEFSMERSSENTYKSRRRSYHSSLRLSAHRLNSDSGHKSDTHRSGGRERGRSKERKHLLSPDVSRCNSEERGTQADWESPERRQSRSPSEGRSQTPNRQGTGSLSESSIPSVSDTSTPRRSRRQLPPVPPKPRPLLSYSSLIRHAGSISPPADGSEEGSPLTSQALESNNACLTESSNSPHPQQSQHASPQRYISEPYLALHEDSHASDCGEEETLTFEAAVATSLGRSNTIGSAPPLRHSWQMPNGHYRRRRRGGPGPGMMCGAVNNLLSDTEEDDKC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title